Unidades de filtro para centrífuga, Vivaspin® 2
Proveedor: Sartorius
VS0211 VS0221 VS0231 VS0241 VS02V2 VS0212 VS0202 VS0222 VS0232 VS0242 VS0291 VS0292 VS02H01 VS02H02 VS02H11 VS02H12 VS02H21 VS02H22 VS0201 VS02H91 VS02H92
512-3753EA 204 EUR
512-3753 512-3754 512-3755 512-3756 512-3772 512-4006 512-4007 512-4008 512-4009 512-4010 512-2841 512-2842 512-2843 512-2844 512-2845 512-2846 512-2847 512-2848 512-3752 512-2866 512-2867
Unidades de filtro para centrífuga, Vivaspin® 2
Filtros Filtros centrífugos
Vivaspin® concentrators are disposable ultrafiltration devices for the concentration of biological samples of 0,4 to 2 ml.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
Vivaspin® 2 can effectively be used in swing bucket or fixed angle rotors accepting 15 ml centrifuge tubes. The patented vertical membrane design and thin channel filtration chamber (US 5,647,990) minimisesMore Product Information