Quantitative filter papers, ashless grades, grade 589/3, Whatman™
Proveedor: Whatman products (Cytiva)
10300210 10300211 10300212 10300214 10300263 10300213
512-3613EA 43.58 EUR
512-3613 512-3614 512-3615 512-3616 512-0402 WHAT10300213
Quantitative filter papers, ashless grades, grade 589/3, Whatman™
Filtros Papeles de filtros Papel de filtro cuantitativo
Whatman Grade 589/3 ashless filter paper from Cytiva is particularly useful in capturing very fine particulate samples such as insoluble molecules in fats or oils or supporting ash free dry mass.
  • High purity (<0,01% ash) for ash free dry mass methods
  • Blue ribbon filter
  • Very fine precipitate retention (class 2d acc. to DIN 53 135)
  • Herzberg filtration speed of 750 s*
  • Also available prepleated as Grade 589/3 ½
*The time taken to filter 100 ml water (20° C) through a filter area of 10 cm² at a constant pressure of 5 cm water column.
Grade 589/3More Product Information