Glass microfibre filters for personal air samplers, grade GF/A, Whatman™
Proveedor: Whatman products (Cytiva)
18208296
513-0105EA 144.92 EUR
513-0105
Glass microfibre filters for personal air samplers, grade GF/A, Whatman™
Filtros Filtros de fibras de vidrio
Grade GF/A card-mounted filters are used in static sample and personal air sampler applications. These aerosol sampling and particulate monitoring filters provide high flow rates and minimal sample interference.
  • Heat resistant up to 500 °C
  • Made of high quality borosilicate glass fibres without added binders
  • Low moisture affinity and high loading capacity
  • Card mounted for use with a continuous air monitor or personal air sampler
Accurate suspended particulate matter analysis is critical in nuclear plants and similar environments.
The GF/A cardMore Product Information