Filtros en línea, ReZist™, Whatman™
Proveedor: Whatman products (Cytiva)
10463607 10463608 10463609
514-0090EA 170 EUR
514-0090 514-0091 514-0092
Filtros en línea, ReZist™, Whatman™
Filtros Filtros en línea
These in-line filters with PTFE membrane and PP housing are lightweight air venting units suitable for protective vents, in-line filtration and isolation applications when working with chemically harsh solvents.
  • Integral hydrophobic PTFE venting membrane is resistant to harsh solvents
  • Can be autoclaved multiple times without loss of integrity
  • Integrity testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight: avoids the collapsing of tubing that can be caused by heavy filter devices
  • Appropriate for in-line HPLC sample preparation