VWR® Tube Top Vacuum Filtration Systems
Proveedor: VWR Collection
514-1068EA 240 EUR
514-1068 514-1069 514-1070 514-1071
VWR® Tube Top Vacuum Filtration Systems
Filtros Filtros de la parte superior de botellas
These units are ideal for minimising unnecessary transfers by filtering directly into the centrifuge tube.
  • Sterilised by electron beam irradiation
  • Non pyrogenic
PES membrane: Low protein binding and low extractables. Ideal for tissue culture applications.
Información de suministro: Each PP centrifuge tube is supplied with an individually wrapped cap for storage. Each individual unit is lot-numbered for easy identification and tracking.
Envase: Packaged in resealable bag. Centrifuge tube cap is individually wrapped.
Precaución: Funnel/tube size.
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental ISO 14001/Certificate No.: 04923E00243R0M