Clarification and sterile filtration kit, Sartoclear Dynamics® Lab P15
Proveedor: Sartorius
SDLP-0015-C
514-1073EA 163 EUR
514-1073
Clarification and sterile filtration kit, Sartoclear Dynamics® Lab P15
Filtros Filtros de jeringas
Sartoclear Dynamics® Lab products are filtration kits designed for the clarification and sterile filtration of mammalian cell cultures in R and D laboratories. The Sartoclear Dynamics® P15 kit is designed for the clarification and sterile filtration of 15 ml mammalian cell culture.
  • Ready to use
  • Fast and effortless
  • Clarification technology available for laboratories but also for process volumes
Sartoclear Dynamics® Lab kits enable clarification and sterile filtration to be performed in a single step. These kits simplify the process by fully eliminatingMore Product Information