Slide-A-Lyzer™ G3 Dialysis Cassettes
Proveedor: Thermo Fisher Scientific
A52966 A52971 A52961 A52962 A52963 A52964 A52965 A52967 A52968 A52969 A52970 A52972 A52973 A52974 A52975 A52976 A52977 A52978 A52979 A52980 A52981 A52982 A52983 A52984 A52985
514-1383EA 141 EUR
514-1383 514-1388 514-1378 514-1379 514-1380 514-1381 514-1382 514-1384 514-1385 514-1386 514-1387 514-1389 514-1390 514-1391 514-1392 514-1393 514-1394 514-1395 514-1396 514-1397 514-1398 514-1399 514-1400 514-1401 514-1402
Slide-A-Lyzer™ G3 Dialysis Cassettes
Tuberías Tuberías para diálisis
Thermo Scientific Slide-A-Lyzer™ G3 Dialysis Cassettes facilitate the rapid and trouble-free dialysis of sample volumes from 0,5 to 125 ml. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.
  • Efficient dialysis - flat cassette chamber, housing two membranes, provides high surface area to volume ratio maximising diffusion rate
  • Ease-of-use - sample loading and removal with a pipette and syringe
  • High sample recovery - low binding plastic and membrane help minimise sample loss compared