Mixed cellulose ester membranes, non-sterile, 0,45 µm, Whatman™
Proveedor: Whatman products (Cytiva)
7141-004 10401606 10401612 10401614 10401618 10401621 10401626 10401631 10406512 10406812 10406814 10409414 10409714 10401664 7141-204 66191ME
516-1144EA 201 EUR
516-1144 514-2151 514-2152 514-2153 514-0099 514-2155 514-0763 514-2154 514-3302 514-3342 514-3352 514-3393 514-3384 514-0944 514-0543 514-1448
Mixed cellulose ester membranes, non-sterile, 0,45 µm, Whatman™
Filtros Filtros de membranas
Mixed cellulose ester, WME range, circles, gridded. Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high