Capsule filters, Polycap™ SPF, Whatman™
Proveedor: Whatman products (Cytiva)
6705-3600 6705-7500
515-0107EA 84.78 EUR
515-0107 515-0110
Capsule filters, Polycap™ SPF, Whatman™
Filtros Cápsulas de filtro de membrana
These Polycap™ SPF capsule filters are optimised for prefiltration applications. They are typically used upstream of a Polycap™ AS or Polycap™ TC capsule during enzyme, nutrient media or buffer filtration.
  • Three layers of filter media: fine glass microfiber (GMF), ultrafine GMF, and polyethersulfone (PES)
  • Suitable for hard-to-filter solutions, particularly sera and protein solutions
  • Can be sterilized by autoclaving
  • Well suited for serum, viral suspension, nutrient, biological, immunological, enzyme, and buffer filtration
Biological filtration,More Product Information