Qualitative filter papers, standard grades, grade 597 and 597¹/₂, Whatman™
Proveedor: Whatman products (Cytiva)
10311804 10311807 10311808 10311809 10311810 10311811 10311812 10311814 10311820 10311822 10311841 10311842 10311843 10311844 10311845 10311847 10311851 10311852 10311853 10311854 10311856 10311862 10311887 10311897
515-4299EA 7.64 EUR
515-4299 515-4300 515-4301 515-4302 515-4303 515-4304 515-4305 515-4306 515-4307 515-4309 515-5301 515-5302 515-5303 512-3628 512-3629 512-3630 512-3631 512-3632 512-3633 512-3634 515-5311 512-0004 512-0577 515-4313
Qualitative filter papers, standard grades, grade 597 and 597¹/₂, Whatman™
Filtros Papeles de filtros Papel de filtro cualitativo
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
  • Increased total loading capacity as more filter area is available
Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
The prepleating does not significantly affect any of the technicalMore Product Information