Centrifugal filter units, Amicon® Ultra-4
Proveedor: Merck
UFC801024 UFC801096 UFC803024 UFC803096 UFC810024 UFC810096 UFC801008 UFC805024 UFC803008 UFC805008 UFC810008 UFC800324 UFC800396 UFC805096 UFC800308
516-0553EA 277 EUR
516-0553 MILFUFC801096 516-0554 MILFUFC803096 516-0624 MILFUFC810096 516-0619 516-0622 516-0620 516-0621 516-0623 MILFUFC800324 MILFUFC800396 MILFUFC805096 MILFUFC800308
Centrifugal filter units, Amicon® Ultra-4
Filtros Filtros centrífugos
These centrifugal filter units can be used for a number of applications: concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; purification of macromolecular components found in tissue culture extracts or cell lysates and protein removal prior to HPLC; concentration of dilute or pre-purified proteins from column eluents; desalting, buffer exchange and protein dialysis; size exclusion and protein fractionation.
  • High retentate recovery of >90%; 100% integrity tested
  • Vertical membrane reduces