Ashless hardened quantitative filter paper
Proveedor: Ahlstrom-Munksjö
103125 103118 103114 103035 103167 103028 103032 103025 103100 103111 103332 103171 103179 103107 103233 103023 103038 103040 103042 103103 103123 103162 103163 103173 103182 103184 103222 103226 103229 103235 103239 103242 103244 103282 103283 103285 103288 103291 103294 103295 103297 103329 103331 103333 103335 103251 103304 103009 103076 103149 103211 103049 103252 103321 103310 103309 103305 103253 103250 103189 103141 103138 103136 103132 103056 103255 103096 103190 103020 103021 103097 103128 103157 103159 103186 103220 103221 103247 103280 103281 103301 103271 103044 103348 103046 103191 103193 103197 103328 103339 103340 103343 103344 103345 103346
516-0711EA 128 EUR
516-0711 516-0709 516-0708 516-0698 516-0717 516-0696 516-0697 516-0695 516-0704 516-0707 516-1075 516-0718 516-0720 516-0706 516-0729 516-0694 516-0699 516-0700 516-0701 516-0705 516-0710 516-0715 516-0716 516-0719 516-0721 516-0722 516-0726 516-0727 516-0728 516-0730 516-0731 516-0732 516-0733 516-0737 516-0738 516-0739 516-0740 516-0741 516-0742 516-0743 516-0744 516-1073 516-1074 516-1076 516-1077 516-1062 516-1067 516-1045 516-1049 516-1054 516-1060 516-1047 516-1063 516-1071 516-1070 516-1069 516-1068 516-1064 516-1061 516-1055 516-1053 516-1052 516-1051 516-1050 516-1048 516-1065 516-0702 516-1056 516-0692 516-0693 516-0703 516-0712 516-0713 516-0714 516-0723 516-0724 516-0725 516-0734 516-0735 516-0736 516-0745 516-1066 516-0900 516-1084 516-1046 516-1057 516-1058 516-1059 516-1072 516-1078 516-1079 516-1080 516-1081 516-1082 516-1083
Ashless hardened quantitative filter paper
Filtros Papeles de filtros Papel de filtro cuantitativo
Ahlstrom-Munksjö manufactures a series of extremely high purity, acid washed filter papers designed for use in analytical and gravimetric analysis. The quantitative grades are manufactured from top quality cotton linters using ultra-pure, deionised water and are further treated with dilute acid to remove any remaining organic and inorganic impurities. All quantitative grades are manufactured in a strictly controlled environment that ensures high uniformity and high purity from filter to filter.
These filters are treated with a strong acid to remove trace metals and produce high wetMore Product Information