Syringe filters, nylon, Puradisc™, Whatman™
Proveedor: Whatman products (Cytiva)
6750-2502 6750-2504 6751-2502 6751-2504 6786-1301 6789-1301 6789-1302 6789-1304 6790-1302 6790-1304 6750-2510 6752-2504 6753-2502 6753-2504 6753-2510 6768-1302 6768-1304
514-8014EA 192 EUR
514-8014 514-8015 514-8016 514-8017 516-0925 516-0436 516-1271 516-1273 516-1272 516-1274 514-0228 514-0230 514-0231 514-0232 514-0233 516-0922 516-0923
Syringe filters, nylon, Puradisc™, Whatman™
Filtros Filtros de jeringas
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample typesMore Product Information