Millex® Automation-Compatible Filter Units, with Hydrophilic LCR (PTFE) Membrane
Proveedor: Merck
SLCRDZ5NK SLCRBZ5NK SLCRBZ5NZ SLCRDZ5NZ SLLGDZ5NK SLLGDZ5NZ
516-6553EA 2690 EUR
516-6553 MILFSLCRBZ5NK MILFSLCRBZ5NZ MILFSLCRDZ5NZ MILFSLLGDZ5NK MILFSLLGDZ5NZ
Millex® Automation-Compatible Filter Units, with Hydrophilic LCR (PTFE) Membrane
Filtros Filtros de jeringas
Automation compatible filter units are specially designed to provide efficient filtration for dissolution and other automated sample preparation applications.
  • Domed housing ensures reliable delivery of filters
  • Pressure-resistant housing resists bursting
  • Luer-Lok® connection optimised for precise alignment and fit
Automation compatible filter units are available in a variety of membrane types and pore sizes, including units with pre-filters for particulate-laden samples. Used in the process of drug dissolution testing, automated sample preparation,More Product Information