Bioprocessing centrifuges, refrigerated, floor standing, Thermo Scientific™ Sorvall™ LYNX™
Proveedor: Thermo Fisher Scientific
75006580 75006590 75006591 75006581
521-1979EA 42800 EUR
521-1979 521-1980 521-2684 521-2683
Bioprocessing centrifuges, refrigerated, floor standing, Thermo Scientific™ Sorvall™ LYNX™
Centrífugas Centrífugas de suelo
These bioprocessing centrifuges offer exceptional performance and large capacities up to 4 or 6 litres to meet the evolving application needs of production facilities. Ideal for bioprocessing and drug discovery.
  • Auto-ID instant rotor identification: Immediate identification of a rotor when secured in the centrifuge chamber, with rotor specifications automatically loaded into the centrifuge parameters
  • Auto-lock rotor exchange: Secure, push-button rotor exchange in as little as 3 seconds
  • Touch screen interface: Simple set-up with large, bright interactive