Phosphate buffered saline, PBS-TWEEN® tablets, Millipore®
Proveedor: Merck Millipore (Oncogene)
524653-1
524653-1EA 97.5 EUR
524653-1
Phosphate buffered saline, PBS-TWEEN® tablets, Millipore®
Tampones Tampones para cultivo celular
Developed for use in immunoassay applications.
One tablet dissolved in 1 litre of deionised water yields 140 mM NaCl, 3 mM KCl, 0,05% TWEEN 20 detergent, 10 mM phosphate buffer with a pH of 7,4 at 25 °C.
Can be stored at 15 to 30 °C.