Vacuum concentrator, DNA SpeedVac system
Proveedor: THERMO SAVANT
DNA130-230
531-1493EA 6720 EUR
531-1493
Vacuum concentrator, DNA SpeedVac system
Concentradores
Compact and integrated vacuum concentrator for sample safety and reproducible results.
  • Compact, easy to use design
  • Lid lock illuminates when lid is closed and locked
  • External post trap neutralizes ammonium hydroxide vapors
  • Data integrity and transfer ability to download data from a USB port
  • Modes of operation: Auto, manual and pre-heat temperature
Designed for DNA and RNA applications, the compact and integrated Thermo Scientific™ Savant™ SpeedVac™ DNA130 vacuum concentrator supports the rapid and safe concentration and drying-down of samples,More Product Information