Rotary Evaporators, Hei-VAP Industrial B
Proveedor: Heidolph Instruments GmbH & Co.KG
518-21100-00 518-22100-00 518-23100-00 518-24100-00 518-25100-00 518-26100-00 518-28100-00 518-28102-00
531-3431EA 0 EUR
531-3431 531-3432 531-3433 531-3434 531-3435 531-3436 531-3106 HEID518-28102-00
Rotary Evaporators, Hei-VAP Industrial B
Evaporadores Rotavapores
Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180 °C. Receiver cassettes protect against threat of glassware breakage.
  • Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
  • Optional Base Cart is fully-maneuverable with removable panels for storage of vacuum pump, glassware, solvents or accessories
  • Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
  • Large touch screen control