Conos de sedimentación Imhoff
Proveedor: VITLAB
75991
532-0099EA 39.1 EUR
532-0099
Conos de sedimentación Imhoff
Embudos Conos de sedimentación
Styrene-acrylonitrile (SAN), PP screw plug.
  • As transparent as glass
  • Easy draining via screw plug
Graduation: 0 to 2 ml:0,1/ 2 to 10 ml:0,5/ 10 to 40 ml:1/ 40 to 100 ml:2/ 100 to 1000 ml:10.