Supelco® TraceCERT® Organic Reference Standard, Chlordane (mixture of isomers)
Proveedor: Merck
53797-20MG
53797-20MGEA 196 EUR
53797-20MG
Supelco® TraceCERT® Organic Reference Standard, Chlordane (mixture of isomers)
Normativas Estándares orgánicos
Certificaciones: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Precaución: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312