Scintillation vials, PET, WHEATON®
Proveedor: DWK Life Sciences
986754 986734 986736 986740 986741 986742 986744 986746 986750 986751 986756 986752 986731 986730 986732
WHEA986754EA 372 EUR
WHEA986754 548-2048 548-2049 WHEA986740 WHEA986741 WHEA986742 WHEA986744 WHEA986746 WHEA986750 WHEA986751 WHEA986756 WHEA986752 548-2047 548-2045 548-2046
Scintillation vials, PET, WHEATON®
Viales
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
  • Clarity of glass
  • Can be safely incinerated
  • Lightweight and virtually unbreakable
* With cap on