VWR®, Microjeringa para volumen muy reducido
Proveedor: VWR Collection
549-0198EA 53.6 EUR
549-0198 549-0197
VWR®, Microjeringa para volumen muy reducido
Jeringas Jeringas para cromatografía
Micro syringe with fixed needle for high sensitivity GC analysis and ultra-minute volumes.
  • Zero dead volume between plunger and needle
  • Fixed needle with bevelled tip for smooth septum penetration
Length in ordering table refers to needle length.