Protein based chiral HPLC columns, CHIRALPAK® HSA
Proveedor: CHIRAL TECHNOLOGIES
34714 34794 34793 34792 34791 34783 34782 34781 34733 34713 34712 34711 34734 34784
554-2496EA 1790 EUR
554-2496 554-2514 554-2513 554-2512 554-2511 554-2499 554-2498 554-2497 554-3496 554-2495 554-2494 554-3498 554-3497 554-2510
Protein based chiral HPLC columns, CHIRALPAK® HSA
Columnas de cromatografía
These protein-based columns are suitable for separation of acidic and non-protolytic compounds. These columns function entirely in the reversed phase chromatographic mode, using buffers with low organic modifier content and at moderate pH values.
CHIRALPAK® HSA is narrower applicability than CHIRALPAK® AGP. Separates preferentially weak and strong acids and non-protolytic compounds.
The chiral selector in this stationary phase is human serum albumin (HSA). The protein has been immobilised onto spherical 5 μm particles. Enantiomers of preferentially acidic compounds can be resolvedMore Product Information