Columnas de fase reversa del HPLC, PRP-1
Proveedor: HAMILTON BONADUZ
79444 79753 79790 79366 79443 79479 79820 79558 79423 79571 79755 79529 79422 79380 79427 79351 79381 79795 79531 79352 79495 79499 79496 79478 79567 79428 79493 79525 79447 79317 79445 79318 79121 79122 79425 79426 79355 79367 79229 79154 79286 79912
554-1259EA 917 EUR
554-1259 554-0431 554-0466 554-0211 554-0245 554-0258 554-0484 554-1308 554-0230 554-1313 554-0433 554-0294 554-0229 554-0213 554-0233 554-0202 554-0214 554-0468 554-0295 554-0203 554-0285 554-0289 554-0286 554-0257 554-1312 554-0234 554-0284 554-0290 554-1262 554-1240 554-1260 554-1241 554-1221 554-1222 554-0231 554-0232 554-2537 554-1494 554-2536 554-2531 554-5698 554-5700
Columnas de fase reversa del HPLC, PRP-1
Columnas de cromatografía
PRP-1 polymeric HPLC columns are designed for broad ranging uses, similar to C8 or C18 HPLC materials. They combine the best characteristics of silica-based and polymeric columns resulting in highly inert and long-lasting columns. These columns are ideal for applications that require a high degree of column to column and batch to batch consistency (quality assurance, clinical trials). Unlike C8 or C18 columns, PRP-1 has no stationary phase coating. Since there is no stationary phase to hydrolyse, the column maintains its performance characteristics longer than many C8 or C18 columns.