HPLC columns, Acclaim™ Carbamate
Proveedor: Thermo Fisher Scientific
072926
555-7484EA 1070 EUR
555-7484
HPLC columns, Acclaim™ Carbamate
Columnas de cromatografía
Columns are used for the separation of carbamate pesticides.
  • Compatible with both binary and ternary mobile phase gradients
  • High efficiency
  • Low column bleed
  • Rugged column packing