HPLC columns, Acclaim™ RSLC Explosives E2
Proveedor: Thermo Fisher Scientific
076226
555-7512EA 1090 EUR
555-7512
HPLC columns, Acclaim™ RSLC Explosives E2
Columnas de cromatografía
Acclaim explosives columns are specifically designed for explosives analysis (EPA Method 8330) and are ideal for nitroaromatics and nitramines.
  • Reproducible performance
  • Simple isocratic elution conditions
  • Unique selectivities for separating other nitroaromatic molecules
  • Rugged columns with good lot-to-lot reproducibility