GC capillary column connectors, SilTite µ-Union
Proveedor: Thermo Fisher Scientific
290SM301 290SM302 290SM303 290SM304 290SM305 290SM321 290SM322 290SM401 290SM402 290SM403 290SM404 290SM405
555-9136EA 191 EUR
555-9136 555-9135 555-9134 555-9133 555-9132 555-9192 555-9191 555-9189 555-9188 555-9187 555-9186 555-9185
GC capillary column connectors, SilTite µ-Union
Racores y conectores
The SilTite µ-Union is a connector for GC capillary columns, giving zero dead volume. It has low thermal mass, is only 9 mm in length and has a mass <0,5 g.
  • Zero dead volume giving optimised peak shapes
  • FingerTight technology for leak free installation
  • Highly inert and robust
Información de suministro: Each SilTite µ-Union kit contains: 5× ferrules, 2× union fittings, installation jigs, installation instructions.