Columnas capilares, BPX608
Proveedor: Trajan Scientific and Medical
054823
568-0461EA 874 EUR
568-0461
Columnas capilares, BPX608
Columnas de cromatografía
These columns are optimised for ECD. Ideal for organochlorine, pesticides and herbicides analysis.
35% phenyl polysilphenylene-siloxane
Max. temperature: 10 to 360/370 °C
Suitable replacement for: DB-608, Rtx-35, SPB-608, HP-35, ZB-35.