HPLC columns for carbohydrate analysis, Hi-Plex
Proveedor: VARIAN
PL1F70-6850 PL1170-1810 PL1170-1820 PL1170-1830 PL1170-1840 PL1170-1850 PL1170-1860 PL1170-2820 PL1170-2823 PL1170-6810 PL1170-6820 PL1170-6830 PL1170-6840 PL1170-6860 PL1171-1140 PL1F70-6830 PL1570-5810 PL1670-0810 PL1670-0820 PL1670-0830 PL1670-0840 PL1670-0850 PL1670-0860 PL1671-0140 PL1171-6140
568-0840EA 2420 EUR
568-0840 VARIPL1170-1810 VARIPL1170-1820 VARIPL1170-1830 VARIPL1170-1840 VARIPL1170-1850 VARIPL1170-1860 568-0790 568-0791 568-0792 568-0793 568-0794 568-0795 568-0796 VARIPL1171-1140 568-0839 568-0819 VARIPL1670-0810 VARIPL1670-0820 VARIPL1670-0830 VARIPL1670-0840 VARIPL1670-0850 VARIPL1670-0860 VARIPL1671-0140 568-0797
HPLC columns for carbohydrate analysis, Hi-Plex
Columnas de cromatografía
Hi-Plex HPLC columns deliver improved efficiency, lower operating pressures and longer column lifetimes from monodispersed materials. With a range of ligand counter ions for optimum selectivity and resolution and materials matched to the USP definitions of media types L17, L19, L34 and L58, the Hi-Plex range is ideal for isocratic separations using water or dilute acid as the eluent. This simplifies your HPLC system requirements and eliminates the use of potentially hazardous organic solvents.
  • Simplified HPLC system requirements through isocratic separation capabilities
  • Can be used