HPLC columns, PLRP-S
Proveedor: VARIAN
PL1912-3802 PL1512-5500 PL1912-1501 PL1912-1502 PL1512-5802 PL1111-3500 PL1912-1300 PL1912-1301 PL1912-1500 PL1912-1503 PL1912-1801 PL1912-1802 PL1912-1803 PL1912-3300 PL1912-3301 PL1912-3500 PL1912-3501 PL1912-3801 PL1912-3803 PL1912-5500 PL1912-5501 PL1912-5801 PL1512-1300 PL1512-1301 PL1512-1500 PL1512-1501 PL1512-1502 PL1512-1503 PL1512-1801 PL1512-1802 PL1512-1803 PL1512-3300 PL1512-3301 PL1512-3501 PL1512-3800 PL1512-3801 PL1512-3802 PL1512-3803 PL1512-5501 PL1512-5800 PL1512-5801
568-0834EA 1160 EUR
568-0834 568-0814 568-0823 568-0824 568-0818 568-0789 568-0820 568-0821 568-0822 568-0825 568-0826 568-0827 568-0828 568-0829 568-0830 568-0831 568-0832 568-0833 568-0835 568-0836 568-0837 568-0838 568-0798 568-0799 568-0800 568-0801 568-0802 568-0803 568-0804 568-0805 568-0806 568-0807 568-0808 568-0809 568-0810 568-0811 568-0812 568-0813 568-0815 568-0816 568-0817
HPLC columns, PLRP-S
Columnas de cromatografía
The PLRP-S family of products consists of a range of pore sizes and particle sizes, all with identical chemistry and fundamental adsorptive characteristics. The particles are inherently hydrophobic so there is no bonded phase, alkyl ligand required for reversed-phase separations. This gives a highly reproducible material that is free from silanols and heavy metal ions. Within the extensive product range are columns suitable for analytical separations and preparative purifications.
  • Contain durable and resilient particles that deliver reproducible results