Supelco® Ascentis® Silica HPLC Columns
Proveedor: Merck
581500-U 581502-U 581503-U 581509-U 581510-U 581512-U 581513-U 581514-U 581515-U 581518-U 581519-U 581522-U
581500-UEA 550 EUR
581500-U 581502-U 581503-U 581509-U 581510-U 581512-U 581513-U 581514-U 581515-U 581518-U 581519-U 581522-U
Supelco® Ascentis® Silica HPLC Columns
Columnas de cromatografía
Ascentis® Silica HPLC columns for the classic use in normal phase HPLC.
  • High purity, type B silica for inertness, reproducibility, and stability
  • Modern bonding process optimises bonded phase coverage and maximises stability, while minimizing bleed and unwanted secondary interactions
  • Compatible with LC-MS and all of today's sensitive instruments and methods
  • Scalable sensitivity from analytical to preparative
  • High surface area silica for high preparative loading capacity
  • Impurity: <5 ppm metals
The right structure of the silica surface, as opposed toMore Product Information