Canisters, GL 45, b.safe
Proveedor: Bohlender
R205-02 R205-05 R205-10
590-0287EA 14.4 EUR
590-0287 590-0288 590-0289
Canisters, GL 45, b.safe
Contenedores
Canister with thread GL 45 (DIN 45) for the collection of liquid waste.
  • Including screw cap
  • With UN-approval
  • b.safe accessories such as collecting trays, spouts and grounding cables, for dissipative canisters available
b.safe offers also suitable containers like flasks and canisters in addition to the wide range of caps for the extraction of solvents and waste caps or funnels for the collection of liquid waste.
Unless otherwise indicated, all b.safe canisters are supplied with UN-approval and can be used for the transport of hazardous goods.