SafetyWasteCaps
Proveedor: S.C.A.T.
107949 107960 107962 107963 107966 107967 107969 108023 108024 108025 108026 108046 108047 108156 108200 108201 108205 108231 107034 107036 107037 107038 107029 107913 107915 107917 107922 107924 107926 107927 107928 107930 107935 107936 107942 107943 107947 107050 108055 107053 107052 107051 107257 107256 107259 107242 107246 108142 307912 307923 308921 307964 307500 308964 307925 307931 307944 308403 307916 307918 308401 308402 308404 308961 307108 307109 307961 107121 108143
590-0646EA 195 EUR
590-0646 590-0657 590-0659 590-0660 590-0663 590-0664 590-0666 590-0713 590-0714 590-0715 590-0716 590-0735 590-0736 590-0765 590-0766 590-0767 590-0771 590-0774 590-0520 590-0522 590-0523 590-0524 590-0515 590-0618 590-0620 590-0622 590-0626 590-0628 590-0630 590-0631 590-0632 590-0634 590-0635 590-0636 590-0640 590-0641 590-0645 554-1546 SCTV108055 SCTV107053 SCTV107052 SCTV107051 SCTV107257 SCTV107256 SCTV107259 SCTV107242 SCTV107246 SCTV108142 SCTV307912 SCTV307923 SCTV308921 SCTV307964 SCTV307500 SCTV308964 SCTV307925 SCTV307931 SCTV307944 SCTV308403 SCTV307916 SCTV307918 SCTV308401 SCTV308402 SCTV308404 SCTV308961 SCTV307108 SCTV307109 SCTV307961 SCTV107121 SCTV108143
SafetyWasteCaps
Tapas Tapones de seguridad y para residuos
The SCAT SafetyWasteCaps provide maximum safety for your HPLC liquid disposal. Safely dispose of your liquid laboratory waste without contaminating the ambient air by using SafetyWasteCaps with integrated exhaust air filters. These high-performance filters regulate the air pressure balance and prevent the escape of vapors due to the resulting overpressure in the disposal container. The variety of closures, connections and thread sizes allows optimal adaptation to your HPLC situation. Additional versions with grounding connections, level controls or safety funnels or offer every convenienceMore Product Information