HPLC columns, LC-NH₂, SUPELCOSIL, Supelco®
Proveedor: Merck
59132
59132.EA 822 EUR
59132.
HPLC columns, LC-NH₂, SUPELCOSIL, Supelco®
Columnas de cromatografía
The LC-NH2-NP is an amino phase dedicated to normal phase chromatography suitable for separation of wate insoluble analytes like fat soluble vitamins.
  • Show stable retention in normal phase separations
  • Less sensitive to small or varying amounts of water in the mobile phase, relative to unmodified silica
  • Provide excellent separations of fat-soluble vitamins