VWR®, Calibration Weight Sets, Class M1
Proveedor: VWR Collection
611-1854EA 40.8 EUR
611-1854
VWR®, Calibration Weight Sets, Class M1
Pesas Pesas de calibración
  • 10 g stainless steel knob weight
  • 50 g stainless steel knob weight
  • 100 g nickel plated brass knob weight
Información de suministro: Weights are supplied in a bag.