Volumetric flasks, class A, blue markings
Proveedor: SciLabware
FRD202 FRD204 FRD206 FRD208 FRD214 FRD216 FRD218 FRD220 FRD210 FRD212 FRD200 FRD222
612-0079EA 30.6 EUR
612-0079 612-0080 612-0081 612-0082 612-0085 612-0092 612-0093 612-0094 612-3337 612-3719 BIBBFRD200 BIBBFRD222
Volumetric flasks, class A, blue markings
Matraces Matraces aforados
MBL®, borosilicate glass, with smooth neck.
  • Inscriptions in blue enamel
  • Conformity certified, with batch number
ISO 1042
Certificaciones: ISO 1042