VWR®, Slide Boxes with Lid
Proveedor: VWR Collection
612-2706EA 13.3 EUR
612-2706 612-2707 612-2708 631-1517 631-1519 631-1521 631-1522 631-1524 631-1526 631-1506 631-1507 631-1508 631-1510 631-1500 631-1501 631-1502 631-1503 631-1504
VWR®, Slide Boxes with Lid
Archivadores para portaobjetos
Heavy-duty, two-piece microscope slide boxes are designed to store standard slides for transport.
  • Colour coded for easy identification
  • Slide inventory sheet on inside of the cover
  • Separate lid (box for 25 slides)
  • Lid with plastic snap closure (box for 50 slides)
  • Nickel plated clasp and hinge pin (box for 100 slides)
  • Each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box
  • Moulded, rigid box: Securely holds slides in grooved slots
  • Stackable for long-term storage
La caja está fabricada en ABS duradero y aseguradaMore Product Information