Measuring cylinders, tall form, PPCO or PMP, economy, Nalgene®
Proveedor: Thermo Fisher Scientific
3664-0010 3664-0025 3664-0050 3664-0100 3664-0250 3664-0500 3664-1000 3665-0010 3665-0025 3665-0050 3665-0100 3665-0250 3665-0500 3665-1000
NALG3664-0010EA 6.1 EUR
NALG3664-0010 612-2867 612-2868 612-0600 612-2869 612-2870 612-2871 612-2872 612-2873 612-2874 612-2875 612-2876 612-2877 612-2878
Measuring cylinders, tall form, PPCO or PMP, economy, Nalgene®
Probetas Probetas graduadas
Measuring cylinders, made of PPCO, translucent or PMP, clear, tall form.
  • Calibrated “To Contain/To Deliver"
  • No meniscus to confuse readings allow for accurate measurements
  • Easy to read molded graduations
  • Stable with large round base
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Certificación ambiental IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT label IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603)