Matraces aforados, forma trapezoidal, clase A
Proveedor: HIRSCHMANN
2960133 2960140 2960152 2960153 2960160 2960161 2960165 2960170 2960175
612-3441EA 47 EUR
612-3441 612-3442 612-3443 612-3444 612-3445 612-3446 612-3447 612-3448 612-3449
Matraces aforados, forma trapezoidal, clase A
Matraces Matraces aforados
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certificaciones: DIN EN ISO 1042