Graduated pipettes, type 2, Class AS, works certified, MBL
Proveedor: SciLabware
PRA820ASWC PRA822ASWC PRA824ASWC PRA826ASWC PRA828ASWC
612-3960EA 25.8 EUR
612-3960 612-3961 612-3962 612-3963 612-3964
Graduated pipettes, type 2, Class AS, works certified, MBL
Pipetas Pipetas graduadas
Soda lime glass.
  • Calibrated to deliver from any graduation line down to zero at the jet
  • Graduations in permanent amber stain
DIN EN ISO 835
Certificaciones: DIN EN ISO 835.
Información para pedidos: Supplied with calibration certificate showing actual dispensed volume and providing traceability to National Standards.