VWR®, Milk Serological Pipettes
Proveedor: VWR Collection
612-5851EA 95.8 EUR
612-5851 612-5852 612-5853 612-5854
VWR®, Milk Serological Pipettes
Pipetas Pipetas serológicas
PS, transparent, graduated, sterile. These disposable pipettes for liquid transfers are made of GPPS (general polystyrene) with PO (Polyolefin) filter, in compliance with USP Class VI.
  • Temperature resistance: –20 to +50 °C
  • DNase-/RNase-free, non pyrogenic, non cytotoxic
  • Meet the requirement of BSE/TSE, latex-free
  • Sterilised by irradiation
  • Graduations are calibrated for accurate dispensing to within ±2%
  • Every product is printed with a batch number for tracebility
  • Manufactured in a Class 100000 cleanroom environment
  • Manufactured under EN ISO 13485 and ISO 900