Maxitips, Pipette Tips
Proveedor: EPPENDORF
0030048.130 0030050.525 0030050.568
612-7321EA 129 EUR
612-7321 612-7331 612-7333
Maxitips, Pipette Tips
Puntas de pipetas
Eppendorf Maxitips are designed for large volume liquid handling, offering high precision and compatibility with Eppendorf pipetting systems. Ideal for diverse lab applications, they ensure accuracy and ease of use, enhancing workflow efficiency.
  • Use Maxitips P to aspirate 1 to 10 ml from beakers using the positive displacement principle
  • Use Maxitips S to aspirate between 2,5 and 10 ml from high, narrow-mouth vessels with the air cushion principle
  • Valve for Maxitips ensures drip-free dispensing of liquids with a high vapour pressure
  • Graduation on Maxitips