Combitips® advanced Biopur® Individually Blister-Wrapped Positive Displacement Pipette Tips
Proveedor: EPPENDORF
0030089.634 0030089.642 0030089.626 0030089.618 0030089.669 0030089.677 0030089.650 0030089.693 0030089.685
613-2069EA 200 EUR
613-2069 613-2070 613-2068 613-2067 613-2072 613-2073 613-2071 613-2075 613-2074
Combitips® advanced Biopur® Individually Blister-Wrapped Positive Displacement Pipette Tips
Puntas de pipetas
Eppendorf Combitips® advanced, individually blister-wrapped in 'Biopur®' grade, offer unparalleled purity for critical lab tasks, ensuring contamination-free, precise pipetting with an ergonomic design. Ideal for highly sensitive molecular biology applications.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • Nine volume sizes (0,1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters
  • Combitips® advanced in 'Biopur' quality are guaranteed to