Puntas con filtro, ART® Ergonomic
Proveedor: Thermo Fisher Scientific
2330 2350 2330-05 2340-05
736-0208EA 370 EUR
736-0208 736-0210 613-3865 613-3866
Puntas con filtro, ART® Ergonomic
Puntas de pipetas
The unique bi-fabricated wall of the ART® Ergonomic tip creates a strong, effective seal while requiring significantly lower tip insertion and tip ejection force. Available for Gilson Pipetman and FinnPipette Focus + Digital pipettes.
  • Reduce strains and injury
  • Secure and consistent seals
  • Less ejection force
  • Available in low retention polymer
Limited Time Sale
Select items below are currently on sale
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental IRECS; ISO 14001 (Certificate Registration No. EMS 752603)