Bottle-top dispensers, Dispensette® S Organic
Proveedor: Brand
4630130 4630131 4630140 4630141 4630150 4630151 4630160 4630161 4630170 4630171 4630230 4630231 4630240 4630241 4630290 4630291 4630330 4630331 4630340 4630341 4630350 4630351 4630360 4630361
613-5330EA 280 EUR
613-5330 613-5331 613-5332 613-5333 613-5334 613-5335 613-5336 613-5337 613-5338 613-5339 613-5340 613-5341 613-5342 613-5343 613-5344 613-5345 613-5346 613-5347 613-5348 613-5349 613-5350 613-5351 613-5352 613-5353
Bottle-top dispensers, Dispensette® S Organic
Dispensadores de la parte superior del frasco
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Three different models of volume adjustment available: fixed (fixed-volume for standard applications; simple calibration adjustment with supplied tool), analog (analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool) and digital
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for