Matrix™ Reagent Reservoirs
Proveedor: THERMO MATRIX TECHNOLOGIES
8086 8095 8096 8093 8094 8085 8075
MATR8086EA 0 EUR
MATR8086 613-7452 MATR8096 613-7453 613-7451 613-7450 613-5831
Matrix™ Reagent Reservoirs
Depósitos de reactivos
PP or PS. These reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills.
  • 75 ml reservoir made of PP is autoclavable and reusable
  • Trough within a trough maximises the amount of liquid accessible to pipette tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduces spillage when pouring reagents out of the reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags so packaging maintains sterility while being easy to open