Single channel pipettes, mechanical, variable volume, PIPETMAN®
Proveedor: GILSON
F144054M F144055M F144056M F144057M F144058M F144059M F144067 F144066 F167800 F167900 F167360 F144066CG F144067CG F144054MC F144055MC F144066C F144067C F144054MCG F167920
613-5944EA 317 EUR
613-5944 613-5945 613-5946 613-5947 613-5948 613-5949 613-6191 613-5950 613-6500 613-6501 613-5889 GILSF144066CG GILSF144067CG GILSF144054MC GILSF144055MC GILSF144066C GILSF144067C GILSF144054MCG 613-7160
Single channel pipettes, mechanical, variable volume, PIPETMAN®
Pipetas Micropipetas
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling.
  • Legendary performance
  • Easy pipetting
  • Built-to-last and convenient
  • Simple to maintain
PIPETMAN pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting. Eight single channel models cover a volume range from 0.2 µl to 10 ml and eightMore Product Information