epT.I.P.S.® 384 Reloads - Micropipette Tips
Proveedor: EPPENDORF
0030076.001 0030076.010 0030076.044 0030076.052
613-7219EA 463 EUR
613-7219 613-7220 613-7221 613-7222
epT.I.P.S.® 384 Reloads - Micropipette Tips
Puntas de pipetas
Eppendorf epT.I.P.S.® 384 Reloads provide high-precision micropipette tips in 'Eppendorf Quality' and 'PCR clean' grades, tailored for 384-well plate applications, ensuring accuracy, reduced waste, and compatibility in high-throughput labs. The epT.I.P.S.® 384 pipette tips build a system together with the Research® plus and Xplorer® plus 16- and 24-channel pipettes, as well as 20 and 100 µl Move It® adjustable tip spacing pipettes for the utmost safety and reliability.
  • Designed for a cone distance of 4,5 mm
  • Innovative SOFTattach technology allows for