Picus® 2 Single-Channel Pipettes, Electronic
Proveedor: Sartorius
LH-747021 LH-747041 LH-747061 LH-747081 LH-747101 LH-747111
613-7231EA 863 EUR
613-7231 613-7232 613-7233 613-7234 613-7235 613-7236
Picus® 2 Single-Channel Pipettes, Electronic
Pipetas Micropipetas
Picus® 2 pipettes offer reliable, ergonomic pipetting for all users. They feature customisable programs, safety features, and connect to the Sartorius app for automatic adjustments and improved workflow, enhancing laboratory productivity.
  • Reliable and repeatable pipetting results
  • Wide selection of pipetting modes
  • Customisable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app
The Picus® 2 pipettes ensure reliable and repeatable pipetting results with unbeatable ergonomic design that is kind to your hand. Picus® 2 is theMore Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental Renewable energy certificate; ISO 14001