Immersion flow cup
Proveedor: Industrial Physics
VF2071 VF2072 VF2073 VF2074 VF2075 VF2077
616-3412EA 149.3 EUR
616-3412 616-3413 769-1144 616-3414 616-3415 616-3417
Immersion flow cup
Viscosímetros Copas de flujo
Dip cups can be used to provide a quick 'break-thread' flow time comparative measurements on site in a manufacturing environment.
  • Titanium anodised aluminium with fixed stainless steel nozzle and handle
  • Engraved with a unique serial number
  • Easy to clean
Certificaciones: DIN 53211
Información para pedidos: The calibration certificate needs to be ordered separately.
Información de suministro: Stand is not included.